Diphenyl-2-pyridylphosphine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Diphenylphosphino)pyridine
H56029-06
H56029-14
AAH56029-06EA
84.7
USD
AAH56029-06
AAH56029-14
Diphenyl-2-pyridylphosphine 98%
Diphenyl-2-pyridylphosphine
Purity: 98%
CAS No.: 37943-90-1
Molecular Formula: C17H14NP
Formula Weight: 263.27
Melting Point: 82-84°
MDL No.: MFCD00192108
Formula:
C₁₇H₁₄NP Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192108 CAS Number: 37943-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...