Glycine benzyl ester-p-toluenesulfonate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzyl glycinate 4-methylbenzenesulphonate, H-Gly-OBzl.Tos-OH, H-Gly-OBzl.TosOH

H56016-06 H56016-14
AAH56016-06EA 38.9 USD
AAH56016-06 AAH56016-14
Glycine benzyl ester-p-toluenesulfonate 99%
Glycine benzyl ester-p-toluenesulfonate

Purity: 99% CAS No.: 1738-76-7 Molecular Formula: C9H11NO2.C7H8O3S Formula Weight: 337.39 Melting Point: 132-134° Beilstein Reference: 3600116 MDL No.: MFCD00035425

Formula: C₉H₁₁NO₂
MW: 337.39 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00035425
CAS Number: 1738-76-7
EINECS: 217-094-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR