(R)-(+)-2-Bromo-α-methylbenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-(+)-1-(2-Bromophenyl)ethanol, (R)-2-Bromo-alpha-methylbenzyl Alcohol
H56003-03
H56003-06
AAH56003-03EA
55.2
USD
AAH56003-03
AAH56003-06
AAH56003-MD
(R)-(+)-2-Bromo-α-methylbenzyl alcohol 98%
(R)-(+)-2-Bromo-α-methylbenzyl alcohol
Purity: 98%
CAS No.: 76116-20-6
Molecular Formula: C8H9BrO
Formula Weight: 201.06
Melting Point: 54-56°
Optical Rotation: +54° (c=1 in chloroform)
Formula:
C₈H₉BrO MW: 201.06 g/mol Melting Pt: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216591 CAS Number: 76116-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...