2-Acetyl-6-bromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55981-03
H55981-06
AAH55981-03EA
57.3
USD
AAH55981-03
AAH55981-06
AAH55981-MD
2-Acetyl-6-bromopyridine 97%
2-Acetyl-6-bromopyridine
Purity: 97%
CAS No.: 49669-13-8
Molecular Formula: C7H6BrNO
Formula Weight: 200.03
Melting Point: 51-55°
MDL No.: MFCD00272200
Formula:
C₇H₆BrNO MW: 200.03 g/mol Melting Pt: 51…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272200 CAS Number: 49669-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...