3-Amino-1-propanesulfonic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tramiprosate, Homotaurine, 3-Aminopropane-1-sulphonic acid, 3APS

H55918-03 H55918-06 H55918-14
AAH55918-03EA 37.26 USD
AAH55918-03 AAH55918-06 AAH55918-14
3-Amino-1-propanesulfonic acid 97%
3-Amino-1-propanesulphonic acid

Purity: 97% CAS No.: 3687-18-1 Molecular Formula: C3H9NO3S Formula Weight: 139.17 Melting Point: 293° dec MDL No.: MFCD00008225

Formula: C₃H₉NO₃S
MW: 139.17 g/mol
Melting Pt: 293 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008225
CAS Number: 3687-18-1
EINECS: 222-977-4
Merck Index: 14,10218

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR