1-Benzoylpiperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenyl(piperazin-1-yl)methanone

H55896-03 H55896-06
AAH55896-03EA 31.6 USD
AAH55896-03 AAH55896-06 AAH55896-14
1-Benzoylpiperazine 97%
1-Benzoylpiperazine

Purity: 97% CAS No.: 13754-38-6 Molecular Formula: C11H14N2O Formula Weight: 190.24 Melting Point: 61-64° Boiling Point: 145°/0.05mm MDL No.: MFCD00810192

Formula: C₁₁H₁₄N₂O
MW: 190.24 g/mol
Boiling Pt: 145 °C/0.05 mm
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00810192
CAS Number: 13754-38-6
UN: 3263
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR