4-Amino-2-chloropyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chlor-4-pyrimidinamin, 2-Chloro-4-pyrimidinamine, 2-Chloropyrimidin-4-amine, 2-Chloropyrimidin-4-ylamine
H55875-03
H55875-06
AAH55875-03EA
51.9
USD
AAH55875-03
AAH55875-06
AAH55875-MD
4-Amino-2-chloropyrimidine 98%
4-Amino-2-chloropyrimidine
Purity: 98%
CAS No.: 7461-50-9
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Melting Point: 197-205°
MDL No.: MFCD00194051
Formula:
C₄H₄ClN₃ MW: 129.55 g/mol Melting Pt: 197…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00194051 CAS Number: 7461-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...