4,5-Difluoro-2-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Difluoro-6-nitroaniline

H55830-03 H55830-06
AAH55830-03EA 54.5 USD
AAH55830-03 AAH55830-06 AAH55830-MD
4,5-Difluoro-2-nitroaniline 98%
4,5-Difluoro-2-nitroaniline

Purity: 98% CAS No.: 78056-39-0 Molecular Formula: C6H4F2N2O2 Formula Weight: 174.11 Melting Point: 107-108° Beilstein Reference: 2723242

Formula: C₆H₄F₂N₂O₂
MW: 174.11 g/mol
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00010876
CAS Number: 78056-39-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR