2,2'-Dithiodianiline (thulium ionophore I) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Disulphanediyldianiline, 2,2'-Diaminodiphenyl disulphide, 2-Aminophenyl disulphide, Bis(2-aminophenyl) disulphide
H55807-06
H55807-14
AAH55807-03EA
15.13
USD
AAH55807-03
AAH55807-06
AAH55807-14
2,2'-Dithiodianiline (thulium ionophore I) 97%
2,2'-Dithiodianiline (thulium ionophore I)
Purity: 97%
CAS No.: 1141-88-4
Molecular Formula: C12H12N2S2
Formula Weight: 248.36
Melting Point: 91-92°
Beilstein Reference: 914032
Formula:
C₁₂H₁₂N₂S₂ Melting Pt: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007703 CAS Number: 1141-88-4 EINECS: 214-529-1 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...