3-Dodecylthiophene 97%

Supplier: Thermo Scientific Chemicals

H55799-03 H55799-06
AAH55799-03EA 51.8 USD
AAH55799-03 AAH55799-06 AAH55799-14
3-Dodecylthiophene 97%
3-Dodecylthiophene

Purity: 97% CAS No.: 104934-52-3 Molecular Formula: C16H28S Formula Weight: 252.46 Refractive Index: 1.4880 Boiling Point: 290° MDL No.: MFCD00130121

Formula: C₁₆H₂₈S
MW: 252.46 g/mol
Boiling Pt: 290 °C
Density: 0.902
Storage Temperature: Refrigerator
MDL Number: MFCD00130121
CAS Number: 104934-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR