1-(4-Methoxyphenyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H55791-03
H55791-06
H55791-MD
AAH55791-03EA
61
USD
AAH55791-03
AAH55791-06
AAH55791-MD
1-(4-Methoxyphenyl)piperazine 97%
1-(4-Methoxyphenyl)piperazine
Purity: 97%
CAS No.: 38212-30-5
Molecular Formula: C11H16N2O
Formula Weight: 192.26
Melting Point: 42-47°
Beilstein Reference: 156736
Formula:
C₁₁H₁₆N₂O Boiling Pt: 118 °C (0.3 -rr) Melting Pt: 261 - 262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040736 CAS Number: 38212-30-5 EINECS: 253-829-7 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...