2-Chloro-4-hydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H55771-03 H55771-06
AAH55771-03EA 153 USD
AAH55771-03 AAH55771-06 AAH55771-MD
2-Chloro-4-hydroxybenzaldehyde 97%
2-Chloro-4-hydroxybenzaldehyde

Purity: 97% CAS No.: 56962-11-9 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Melting Point: 145-147° MDL No.: MFCD00052184

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00052184
CAS Number: 56962-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR