2,6-Di-tert-butylpyridine 97+%

Supplier: Thermo Scientific Chemicals
Warning

H55761-03 H55761-06 H55761-14
AAH55761-03EA 50.2 USD
AAH55761-03 AAH55761-06 AAH55761-14
2,6-Di-tert-butylpyridine 97+%
2,6-Di-tert-butylpyridine

Purity: 97+% CAS No.: 585-48-8 Molecular Formula: C13H21N Formula Weight: 191.32 Flash Point: 71°(160°F) Refractive Index: 1.4730 Boiling Point: 100-101°/23mm Beilstein Reference: 125886 Merck Reference: 14,3036 MDL No.: MFCD00006306

Formula: C₁₃H₂₁N
Boiling Pt: 100…101 °C/23 mm
Density: 0.867
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6
Merck Index: 14,3036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR