1,10-Phenanthroline-5,6-dione 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55747-03
H55747-06
H55747-MD
AAH55747-03EA
182
USD
AAH55747-03
AAH55747-06
AAH55747-MD
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione
Purity: 98%
CAS No.: 27318-90-7
Molecular Formula: C12H6N2O2
Formula Weight: 210.19
Melting Point: 260° dec.
MDL No.: MFCD00014473
Formula:
C₁₂H₆N₂O₂ Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00014473 CAS Number: 27318-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...