Quinaldine red 95% (dye content)

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

H55729-06 H55729-14
AAH55729-06EA 51.5 USD
AAH55729-06 AAH55729-14
Quinaldine red 95% (dye content)
Quinaldine red

Purity: 95% CAS No.: 117-92-0 Molecular Formula: C21H23IN2 Formula Weight: 430.33 Physical Form: Powder Sensitivity: Light Sensitive Appearance: Green to black Beilstein Reference: 3798055 Merck Reference: 14,8049 MDL No.: MFCD00011968

Formula: C₂₁H₂₃IN₂
MW: 430.33 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00011968
CAS Number: 117-92-0
EINECS: 204-221-5
Merck Index: 12,08230

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR