1,3-Difluoro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Difluoronitrobenzene

H55694-03 H55694-06
AAH55694-03EA 70.8 USD
AAH55694-03 AAH55694-06 AAH55694-MD
1,3-Difluoro-2-nitrobenzene 98%
1,3-Difluoro-2-nitrobenzene

Purity: 98% CAS No.: 19064-24-5 Molecular Formula: C6H3F2NO2 Formula Weight: 159.09 Flash Point: 88°(190°F) Refractive Index: 1.4940 Melting Point: 0° Boiling Point: 91-92°/11mm MDL No.: MFCD00192035

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 91…92 °C/11 mm
Melting Pt: 0 °C
Density: 1.503
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192035
CAS Number: 19064-24-5
EINECS: 242-793-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR