Tris(8-quinolinolato)aluminum 99%

Supplier: Thermo Scientific Chemicals
Warning

H55656-03 H55656-06 H55656-14
AAH55656-03EA 31 USD
AAH55656-03 AAH55656-06 AAH55656-14
Tris(8-quinolinolato)aluminum 99%
Tris(8-quinolinolato)aluminum

Purity: 99% CAS No.: 2085-33-8 Molecular Formula: C27H18AlN3O3 Formula Weight: 459.44 Physical Form: Powder Appearance: Yellow to green Melting Point: 300° MDL No.: MFCD00191693

Formula: C₂₇H₁₈AlN₃O₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191693
CAS Number: 2085-33-8
EINECS: 218-227-0
Merck Index: 14,370

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR