4-Hydroxy-3-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H55597-03 H55597-06
AAH55597-03EA 39.1 USD
AAH55597-03 AAH55597-06 AAH55597-14
4-Hydroxy-3-nitrobenzonitrile 98%
4-Hydroxy-3-nitrobenzonitrile

Purity: 98% CAS No.: 3272-08-0 Molecular Formula: C7H4N2O3 Formula Weight: 164.12 Melting Point: 146-148° Beilstein Reference: 2048724

Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00070775
CAS Number: 3272-08-0
EINECS: 221-899-8
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR