4-Fluoro-2-iodobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

H55531-03 H55531-06
AAH55531-03EA 64.9 USD
AAH55531-03 AAH55531-06 AAH55531-MD
4-Fluoro-2-iodobenzoic acid 97%
4-Fluoro-2-iodobenzoic acid

Purity: 97% CAS No.: 56096-89-0 Molecular Formula: C7H4FIO2 Formula Weight: 266.01 Melting Point: 143-147° UN No.: 2811 MDL No.: MFCD04108278

Formula: C₇H₄FIO₂
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD04108278
CAS Number: 56096-89-0
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR