4-(2-Pyridyl)benzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde
H55498-03
H55498-06
AAH55498-03EA
37.7
USD
AAH55498-03
AAH55498-06
4-(2-Pyridyl)benzaldehyde 98%
4-(2-Pyridyl)benzaldehyde
Purity: 98%
CAS No.: 127406-56-8
Molecular Formula: C12H9NO
Formula Weight: 183.21
Flash Point: 113°(235°F)
Melting Point: 50-52°
MDL No.: MFCD01863537
Formula:
C₁₂H₉NO MW: 183.21 g/mol Melting Pt: 50…52 °C Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01863537 CAS Number: 127406-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...