1,2,3,4-Tetrahydroquinoxaline 98%

Supplier: Thermo Scientific Chemicals
Warning

H55473-03 H55473-06 H55473-MD
AAH55473-03EA 228 USD
AAH55473-03 AAH55473-06 AAH55473-MD
1,2,3,4-Tetrahydroquinoxaline 98%
1,2,3,4-Tetrahydroquinoxaline

Purity: 98% CAS No.: 3476-89-9 Molecular Formula: C8H10N2 Formula Weight: 134.18 Melting Point: 92-98° MDL No.: MFCD00047564

Formula: C₈H₁₀N₂
MW: 134.18 g/mol
Melting Pt: 92…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00047564
CAS Number: 3476-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR