4,6-Dimethyldibenzothiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

H55472-03 H55472-06 H55472-MD
AAH55472-03EA 133 USD
AAH55472-03 AAH55472-06 AAH55472-MD
4,6-Dimethyldibenzothiophene 97%
4,6-Dimethyldibenzothiophene

Purity: 97% CAS No.: 1207-12-1 Molecular Formula: C14H12S Formula Weight: 212.31 Melting Point: 153-157° MDL No.: MFCD00216264

Formula: C₁₄H₁₂S
MW: 212.31 g/mol
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00216264
CAS Number: 1207-12-1
EINECS: 214-894-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR