N,N-Diisopropylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H55471-03 H55471-06
AAH55471-03EA 50.7 USD
AAH55471-03 AAH55471-06 AAH55471-MD
N,N-Diisopropylaniline 97%
N,N-Diisopropylaniline

Purity: 97% CAS No.: 4107-98-6 Molecular Formula: C12H19N Formula Weight: 177.29 Flash Point: 84°(183°F) Physical Form: Liquid Density: 0.91 Refractive Index: 1.5190 Sensitivity: Light Sensitive Appearance: Colorless to pale yellow Boiling Point: 95-96°/11mm MDL No.: MFCD00048277

Formula: C₁₂H₁₉N
Boiling Pt: 95…96 °C/11 mm
Density: 0.91
Flash Pt: 84 °C(183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048277
CAS Number: 4107-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR