4-(4-Pyridyl)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H55462-03 H55462-06
AAH55462-03EA 121 USD
AAH55462-03 AAH55462-06 AAH55462-MD
4-(4-Pyridyl)benzaldehyde 97%
4-(4-Pyridyl)benzaldehyde

Purity: 97% CAS No.: 99163-12-9 Molecular Formula: C12H9NO Formula Weight: 183.21 Melting Point: 85-89° MDL No.: MFCD02684110

Formula: C₁₂H₉NO
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD02684110
CAS Number: 99163-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR