2,5-Dibromohydroquinone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Dibromobenzene-1,4-diol

H55453-03 H55453-06 H55453-14
AAH55453-03EA 24.5 USD
AAH55453-03 AAH55453-06 AAH55453-14
2,5-Dibromohydroquinone 97%
2,5-Dibromohydroquinone

Purity: 97% CAS No.: 14753-51-6 Molecular Formula: C6H4Br2O2 Formula Weight: 267.90 Melting Point: 191-194° MDL No.: MFCD00192664

Formula: C₆H₄Br₂O₂
MW: 267.9 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00192664
CAS Number: 14753-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR