3-Bromo-o-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55436-03
H55436-06
AAH55436-03EA
38.2
USD
AAH55436-03
AAH55436-06
3-Bromo-o-toluic acid 97%
3-Bromo-o-toluic acid
Purity: 97%
CAS No.: 76006-33-2
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 152-156°
MDL No.: MFCD00270097
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00270097 CAS Number: 76006-33-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...