4-Phenylaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl

H55424-03 H55424-06 H55424-14
AAH55424-03EA 24.6 USD
AAH55424-03 AAH55424-06 AAH55424-14
4-Phenylaniline 98%
4-Phenylaniline

Purity: 98% CAS No.: 92-67-1 Molecular Formula: C12H11N Formula Weight: 169.23 Melting Point: 52-54° Boiling Point: 191°/15mm Merck Reference: 14,1235 UN No.: 3077 MDL No.: MFCD00007879

Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 191 °C/15 mm
Melting Pt: 52…54 °C
Density: 1.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007879
CAS Number: 92-67-1
EINECS: 202-177-1
UN: 3077
ADR: 9,III
Merck Index: 12,01276

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR