2,7-Diaminofluorene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55402-03
H55402-06
AAH55402-03EA
97.6
USD
AAH55402-03
AAH55402-06
AAH55402-MD
2,7-Diaminofluorene 97%
2,7-Diaminofluorene
Purity: 97+%
CAS No.: 525-64-4
Molecular Formula: C13H12N2
Formula Weight: 196.25
Melting Point: 160-162°
Beilstein Reference: 2099859
Merck Reference: 14,4156
MDL No.: MFCD00001128
Formula:
C₁₃H₁₂N₂ Boiling Pt: 467 °C (1013 hPa) Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001128 CAS Number: 525-64-4 EINECS: 208-377-5 Merck Index: 13,04182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...