4-Amino-3-iodobenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55400-03
H55400-06
AAH55400-03EA
31.6
USD
AAH55400-03
AAH55400-06
4-Amino-3-iodobenzonitrile 97%
4-Amino-3-iodobenzonitrile
Purity: 97%
CAS No.: 33348-34-4
Molecular Formula: C7H5IN2
Formula Weight: 244.03
Melting Point: 112-115°
MDL No.: MFCD04039964
Formula:
C₇H₅IN₂ Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039964 CAS Number: 33348-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...