4-Bromomethyl-2-cyanobiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

H55394-03 H55394-06 H55394-14
AAH55394-03EA 22.8 USD
AAH55394-03 AAH55394-06 AAH55394-14
4-Bromomethyl-2-cyanobiphenyl 98%
4-Bromomethyl-2-cyanobiphenyl

Purity: 98% CAS No.: 114772-54-2 Molecular Formula: C14H10BrN Formula Weight: 272.14 Melting Point: 125-128° UN No.: 3439 MDL No.: MFCD00671503

Formula: C₁₄H₁₀BrN
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00671503
CAS Number: 114772-54-2
EINECS: 425-280-5
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR