Pyrrole-3-carboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Pyrrole-3-carboxylic acid
H55383-03
H55383-MD
H55383-06
AAH55383-03EA
101
USD
AAH55383-03
AAH55383-MD
AAH55383-06
Pyrrole-3-carboxylic acid 98+%
Pyrrole-3-carboxylic acid
Purity: 98+%
CAS No.: 931-03-3
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Beilstein Reference: 108642
MDL No.: MFCD00800594
Formula:
C₅H₅NO₂ MW: 111.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00800594 CAS Number: 931-03-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...