1,3-Dichloro-2-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

H55365-03 H55365-06 H55365-14
AAH55365-03EA 34.2 USD
AAH55365-03 AAH55365-06 AAH55365-14
1,3-Dichloro-2-iodobenzene 98%
1,3-Dichloro-2-iodobenzene

Purity: 98% CAS No.: 19230-28-5 Molecular Formula: C6H3Cl2I Formula Weight: 272.90 Melting Point: 64-67° MDL No.: MFCD00060658

Formula: C₆H₃Cl₂I
MW: 272.90 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00060658
CAS Number: 19230-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR