Agmatine sulfate 97% (by HPLC)
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate
H55363-03
H55363-06
H55363-MD
AAH55363-03EA
65.14
USD
AAH55363-03
AAH55363-06
AAH55363-MD
Agmatine sulfate 97% (by HPLC)
Agmatine sulfate
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine.
Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.
Formula:
C₅H₁₄N₄ MW: 228.27 g/mol Melting Pt: 231…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013109 CAS Number: 2482-00-0 EINECS: 219-617-3 Merck Index: 14,188 |
Specification Test Results
Assay (HPLC) | >96.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...