trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%

Supplier: Thermo Scientific Chemicals

Synonyms: trans,trans-4-(3,4-Difluorophenyl)-4'-propylbicyclohexyl, trans,trans-4-(3,4-Difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)

H55356-03 H55356-06
AAH55356-03EA 40.1 USD
AAH55356-03 AAH55356-06
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl

Purity: 97% CAS No.: 82832-57-3 Molecular Formula: C21H30F2 Formula Weight: 320.47 Melting Point: 43-46° MDL No.: MFCD09838997

Formula: C₂₁H₃₀F₂
MW: 320.468 g/mol
Melting Pt: 43…46 °C
Storage Temperature: Ambient
MDL Number: MFCD09838997
CAS Number: 82832-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR