4-Methoxysalicylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid

H55345-06 H55345-14
AAH55345-03EA 14.19 USD
AAH55345-03 AAH55345-06 AAH55345-14
4-Methoxysalicylic acid 98%
4-Methoxysalicylic acid

Purity: 98% CAS No.: 2237-36-7 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 158-159° MDL No.: MFCD00002450

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00002450
CAS Number: 2237-36-7
EINECS: 218-801-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR