Di(2-pyridyl) ketone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di(pyridin-2-yl)methanone

H55322-03 H55322-06 H55322-14
AAH55322-03EA 37.4 USD
AAH55322-03 AAH55322-06 AAH55322-14
Di(2-pyridyl) ketone 98%
Di(2-pyridyl) ketone

Purity: 99% CAS No.: 19437-26-4 Molecular Formula: C11H8N2O Formula Weight: 184.20 Flash Point: 110°(230°F) Melting Point: 52-55° Beilstein Reference: 126598 MDL No.: MFCD00006288

Formula: C₁₁H₈N₂O
MW: 184.20 g/mol
Melting Pt: 52…55 °C
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006288
CAS Number: 19437-26-4
EINECS: 243-067-3

Order Now

Specification Test Results

Appearance (Color) White to pale brown
Form Solid
Assay (unspecified) >98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR