3-Bromo-5-fluorophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55288-03
H55288-06
AAH55288-03EA
23.1
USD
AAH55288-03
AAH55288-06
3-Bromo-5-fluorophenol 97%
3-Bromo-5-fluorophenol
Purity: 97%
CAS No.: 433939-27-6
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Flash Point:
Formula:
C₆H₄BrFO Melting Pt: 36…40 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07783710 CAS Number: 433939-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...