4-tert-Butyldimethylsiloxy-1-butanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(tert-Butyldimethylsilyl)oxy-1-butanol, 4-((tert-Butyldimethylsilyl)oxy)butan-1-ol

H55285-06 H55285-14
AAH55285-06EA 49.1 USD
AAH55285-06 AAH55285-14
4-tert-Butyldimethylsiloxy-1-butanol 97%
4-tert-Butyldimethylsiloxy-1-butanol

Purity: 97% CAS No.: 87184-99-4 Molecular Formula: C10H24O2Si Formula Weight: 204.38 Flash Point: 105°(221°F) Refractive Index: 1.4390 Boiling Point: 63-66°/0.3mm MDL No.: MFCD04974070

Formula: C₁₀H₂₄O₂Si
MW: 204.38 g/mol
Boiling Pt: 63…66 °C/0.3 mm
Density: 0.888
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD04974070
CAS Number: 87184-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR