Boc-Cha-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-cyclohexylpropanoic acid, Boc-β-cyclohexyl-Ala-OH
H55282-03
H55282-06
AAH55282-03EA
78
USD
AAH55282-03
AAH55282-06
AAH55282-MD
Boc-Cha-OH 98%
Boc-Cha-OH
Purity: 98%
CAS No.: 37736-82-6
Molecular Formula: C14H25NO4
Formula Weight: 271.36
Beilstein Reference: 3060603
MDL No.: MFCD00211335
Formula:
C₁₄H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00076896 CAS Number: 37736-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...