Potassium tetraphenylborate 97%

Supplier: Thermo Scientific Chemicals
Warning

H55276-06 H55276-14
AAH55276-06EA 62 USD
AAH55276-06 AAH55276-14
Potassium tetraphenylborate 97%
Potassium tetraphenylborate

Purity: 97% CAS No.: 3244-41-5 Molecular Formula: C24H20BK Formula Weight: 358.33 Physical Form: Powder Appearance: White Melting Point: 300° MDL No.: MFCD03093877

Formula: C₂₄H₂₀BK
MW: 358.33 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03093877
CAS Number: 3244-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR