3-Ethynylanisole 96%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
1-Ethynyl-3-methoxybenzene, (3-Methoxyphenyl)ethyne, (3-Methoxyphenyl)acetylene, 1-Methoxy-3-ethynylbenzene
H55254-03
H55254-06
H55254-MD
AAH55254-03EA
54.4
USD
AAH55254-03
AAH55254-06
AAH55254-MD
3-Ethynylanisole 96%
3-Ethynylanisole
Purity: 96%
CAS No.: 768-70-7
Molecular Formula: C9H8O
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.5550
Boiling Point: 204-210°
MDL No.: MFCD00160810
Formula:
C₉H₈O MW: 132.16 g/mol Boiling Pt: 204…210 °C Density: 1.04 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00160810 CAS Number: 768-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...