1-(4-Aminophenyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H55224-03
H55224-06
H55224-MD
AAH55224-03EA
80.5
USD
AAH55224-03
AAH55224-06
AAH55224-MD
1-(4-Aminophenyl)piperazine 97%
1-(4-Aminophenyl)piperazine
Purity: 97%
CAS No.: 67455-41-8
Molecular Formula: C10H15N3
Formula Weight: 177.25
Physical Form: Powder
Appearance: White to purple
Melting Point: 119-123°
MDL No.: MFCD01365906
Formula:
C₁₀H₁₅N₃ Melting Pt: 119…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD01365906 CAS Number: 67455-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...