2-Amino-5-bromobenzonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-2-cyanoaniline
H55217-03
H55217-06
AAH55217-03EA
48.9
USD
AAH55217-03
AAH55217-06
AAH55217-MD
2-Amino-5-bromobenzonitrile 96%
2-Amino-5-bromobenzonitrile
Purity: 96%
CAS No.: 39263-32-6
Molecular Formula: C7H5BrN2
Formula Weight: 197.03
Melting Point: 96-100°
UN No.: 3439
MDL No.: MFCD00158946
Formula:
C₇H₅BrN₂ Melting Pt: 96…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00158946 CAS Number: 39263-32-6 EINECS: 254-387-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...