2-(Methylamino)acetonitrile hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

H55215-06 H55215-14
AAH55215-06EA 38.1 USD
AAH55215-06 AAH55215-14 AAH55215-22
2-(Methylamino)acetonitrile hydrochloride 98%
2-(Methylamino)acetonitrile hydrochloride

Purity: 99% CAS No.: 25808-30-4 Molecular Formula: C3H6N2.HCl Formula Weight: 106.55 Melting Point: 103-106° Beilstein Reference: 3908975 MDL No.: MFCD00012587

Formula: C₃H₇ClN₂
MW: 106.55 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00012587
CAS Number: 25808-30-4
EINECS: 247-277-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR