tert-Butyl (2-bromoethyl)carbamate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(BOC-amino)ethyl bromide

H55211-03 H55211-06
AAH55211-03EA 80.8 USD
AAH55211-03 AAH55211-06 AAH55211-MD
tert-Butyl (2-bromoethyl)carbamate 96%
tert-Butyl (2-bromoethyl)carbamate

Purity: 96% CAS No.: 39684-80-5 Molecular Formula: C7H14BrNO2 Formula Weight: 224.10 Storage Temperature: -30°C to -10°C Melting Point: 30-32° Beilstein Reference: 2325117 MDL No.: MFCD02683428

Formula: C₇H₁₄BrNO₂
MW: 224.10
Melting Pt: 30…32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683428
CAS Number: 39684-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR