3,5-Di-tert-butylpyrocatechol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55207-06
H55207-14
AAH55207-06EA
39.28
USD
AAH55207-06
AAH55207-14
3,5-Di-tert-butylpyrocatechol 99%
3,5-Di-tert-butylpyrocatechol
Purity: 99%
CAS No.: 1020-31-1
Molecular Formula: C14H22O2
Formula Weight: 222.33
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 95-100°
Beilstein Reference: 1370212
MDL No.: MFCD00008819
Formula:
C₁₄H₂₂O₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 95…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008819 CAS Number: 1020-31-1 EINECS: 213-816-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...