1-Methylimidazole-2-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde

H55201-03 H55201-06 H55201-MD
AAH55201-03EA 45.3 USD
AAH55201-03 AAH55201-06 AAH55201-MD
1-Methylimidazole-2-carbaldehyde 98%
1-Methylimidazole-2-carbaldehyde

Purity: 98% CAS No.: 13750-81-7 Molecular Formula: C5H6N2O Formula Weight: 110.12 Flash Point: 102°(216°F) Melting Point: 36-39° Boiling Point: 70-74°/1mm MDL No.: MFCD01321308

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 70…74 °C/1 mm
Melting Pt: 36…39 °C
Flash Pt: 102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD01321308
CAS Number: 13750-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR