(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-4'-(2-Methylbutyl)-[1,1'-biphenyl]-4-carbonitrile
H55183-03
H55183-06
AAH55183-03EA
115
USD
AAH55183-03
AAH55183-06
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 97%
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl
Purity: 97%
CAS No.: 63799-11-1
Molecular Formula: C18H19N
Formula Weight: 249.36
MDL No.: MFCD00192280
Formula:
C₁₈H₁₉N MW: 249.36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00192280 CAS Number: 63799-11-1 EINECS: 264-464-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...