S-Phenyl thioacetate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
H55175-06
H55175-14
AAH55175-06EA
53.5
USD
AAH55175-06
AAH55175-14
S-Phenyl thioacetate 98%
S-Phenyl thioacetate
Purity: 98%
CAS No.: 934-87-2
Molecular Formula: C8H8OS
Formula Weight: 152.21
Flash Point: 79°(174°F)
Physical Form: Liquid
Density: 1.124
Refractive Index: 1.5700
Appearance: Colorless
Boiling Point: 99-100°/6mm
MDL No.: MFCD00008752
Formula:
C₈H₈OS MW: 152.22 g/mol Boiling Pt: 99…100 °C/6 mm Density: 1.124 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008752 CAS Number: 934-87-2 EINECS: 213-294-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...