2-Fluoro-4-nitroaniline 95%

Supplier: Thermo Scientific Chemicals
Warning

H55174-06 H55174-14
AAH55174-06EA 41 USD
AAH55174-06 AAH55174-14 AAH55174-22
2-Fluoro-4-nitroaniline 95%
2-Fluoro-4-nitroaniline

Purity: 95% CAS No.: 369-35-7 Molecular Formula: C6H5FN2O2 Formula Weight: 156.12 Melting Point: 122-130° MDL No.: MFCD00034560

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 122…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00034560
CAS Number: 369-35-7
EINECS: 206-719-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR